Learn More
Cayman Chemical (±)-CP 55,940-d11, Purity: ≥99% deuterated forms (d1-d11), 1 mg
SDP
Catalog No. 502887841
Encompass_Preferred
An internal standard for the quantification of (±)-CP 55,940 by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-288-7841 Supplier Cayman Chemical Supplier No. 107031 MG
An internal standard for the quantification of (±)-CP 55,940 by GC- or LC-MS; intended for research and forensic applications