Learn More
TARGETMOL CHEMICALS INC CP-640186 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T188910MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CP-640186 is an isozyme-nonselective Acetyl-CoA carboxylase (ACCase) inhibitor. The IC50s of CP-640186(CP 640186) is 53 nM and 61 nM for rat liver ACC1 and rat skeletal muscle ACC2 respectively. purity: 98%
Catalog No. 50-258-1066