Learn More
Medchemexpress LLC CP-640186 hydrochloride | 591778-70-0 | C30H36ClN3O3 | 1 ML
SDP
Catalog No. 5000373670
Encompass_Preferred
CP-640186 hydrochloride is an orally active and cell-permeable Acetyl-CoA carboxylase (ACC) inhibitor. It targets ACC1 and ACC2, key enzymes in fatty acid metabolism, and can stimulate muscle fatty acid oxidation.
  • Orally active and cell-permeable
  • Inhibits Acetyl-CoA carboxylase (ACC)
  • Stimulates muscle fatty acid oxidation
  • Purity: 99.65%
  • Solution in DMSO
Catalog No. 50-003-73670 Supplier Medchemexpress LLC Supplier No. HY15259A10MM/1ML
CP-640186 hydrochloride is an orally active and cell-permeable Acetyl-CoA carboxylase (ACC) inhibitor. It targets ACC1 and ACC2, key enzymes in fatty acid metabolism, and can stimulate muscle fatty acid oxidation.
  • Orally active and cell-permeable
  • Inhibits Acetyl-CoA carboxylase (ACC)
  • Stimulates muscle fatty acid oxidation
  • Purity: 99.65%
  • Solution in DMSO