Learn More
Medchemexpress LLC CP-640186 (hydrochloride) | 591778-70-0 | C30H36ClN3O3 | 100 MG
SDP
Catalog No. 5000372305
Encompass_Preferred
CP-640186 hydrochloride is an orally active and cell-permeable inhibitor of Acetyl-CoA carboxylase (ACC). It exhibits IC50 values of 53 nM for rat liver ACC1 and 61 nM for rat skeletal muscle ACC2. ACC is a crucial enzyme involved in fatty acid metabolism, responsible for synthesizing malonyl-CoA. This compound also promotes muscle fatty acid oxidation.
  • Orally active and cell-permeable.
  • Inhibits Acetyl-CoA carboxylase (ACC).
  • IC50s: 53 nM for rat liver ACC1, 61 nM for rat skeletal muscle ACC2.
  • Stimulates muscle fatty acid oxidation.
Catalog No. 50-003-72305 Supplier Medchemexpress LLC Supplier No. HY15259A100MG
CP-640186 hydrochloride is an orally active and cell-permeable inhibitor of Acetyl-CoA carboxylase (ACC). It exhibits IC50 values of 53 nM for rat liver ACC1 and 61 nM for rat skeletal muscle ACC2. ACC is a crucial enzyme involved in fatty acid metabolism, responsible for synthesizing malonyl-CoA. This compound also promotes muscle fatty acid oxidation.
  • Orally active and cell-permeable.
  • Inhibits Acetyl-CoA carboxylase (ACC).
  • IC50s: 53 nM for rat liver ACC1, 61 nM for rat skeletal muscle ACC2.
  • Stimulates muscle fatty acid oxidation.