Learn More
Medchemexpress LLC CP-640186 hydrochloride | 591778-70-0 | C30H36ClN3O3 | 50 MG
SDP
Catalog No. 5000372124
Encompass_Preferred
CP-640186 hydrochloride is an orally active, cell-permeable Acetyl-CoA carboxylase (ACC) inhibitor. It has IC50s of 53 nM for rat liver ACC1 and 61 nM for rat skeletal muscle ACC2. This compound can stimulate muscle fatty acid oxidation and inhibit fatty acid synthesis.
  • Orally active and cell-permeable ACC inhibitor
  • Inhibits rat liver ACC1 and rat skeletal muscle ACC2
  • Stimulates muscle fatty acid oxidation
  • Inhibits H460 cell growth
  • Increases fatty acid metabolism in C2C12 cells and muscle strips
  • Inhibits fatty acid and TG synthesis in HepG2 cells
Catalog No. 50-003-72124 Supplier Medchemexpress LLC Supplier No. HY15259A50MG
CP-640186 hydrochloride is an orally active, cell-permeable Acetyl-CoA carboxylase (ACC) inhibitor. It has IC50s of 53 nM for rat liver ACC1 and 61 nM for rat skeletal muscle ACC2. This compound can stimulate muscle fatty acid oxidation and inhibit fatty acid synthesis.
  • Orally active and cell-permeable ACC inhibitor
  • Inhibits rat liver ACC1 and rat skeletal muscle ACC2
  • Stimulates muscle fatty acid oxidation
  • Inhibits H460 cell growth
  • Increases fatty acid metabolism in C2C12 cells and muscle strips
  • Inhibits fatty acid and TG synthesis in HepG2 cells