Learn More
TARGETMOL CHEMICALS INC CP-673451 10MG
SDP
Catalog No. 502579571
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CP-673451 is a specific inhibitor of PDGFR(alpha)/(beta) (IC50 10/1 nM) with antiangiogenic and antitumor activity and the selectivity is higher 450-fold than other angiogenic receptors. purity: 99%
Catalog No. 50-257-9571 Supplier TARGETMOL CHEMICALS INC Supplier No. T609110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CP-673451 is a specific inhibitor of PDGFR(alpha)/(beta) (IC50 10/1 nM) with antiangiogenic and antitumor activity and the selectivity is higher 450-fold than other angiogenic receptors. purity: 99%