Learn More
Medchemexpress LLC HY-12050 10mg Medchemexpress, CP-673451 CAS:343787-29-1 Purity:>98%
SDP
Catalog No. 502028846
Encompass_Preferred
Medchemexpress, HY-12050 10mg CP-673451 CAS:343787-29-1 CP-673451 is a potent and selective inhibitor of PDGFR with IC50s of 10 and 1 nM for PDGFRα and PDGFRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8846 Supplier Medchemexpress LLC Supplier No. HY1205010MG
Add to Cart
Add to Cart
Medchemexpress, HY-12050 10mg CP-673451 CAS:343787-29-1 CP-673451 is a potent and selective inhibitor of PDGFR with IC50s of 10 and 1 nM for PDGFRα and PDGFRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.