Learn More
Medchemexpress LLC HY-12050 5mg , CP-673451 CAS:343787-29-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120505MG
Encompass_Preferred
HY-12050 5mg CP-673451 CAS: 343787-29-1 CP-673451 is a potent and selective inhibitor of PDGFR with IC50s of 10 and 1 nM for PDGFRα and PDGFRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9649