Learn More
Selleck Chemical LLC CP-673451 5mg 343787-29-1
SDP
Catalog No. NC0521184
Encompass_Preferred
CP-673451 is a selective inhibitor of PDGFR?/? with IC50 of 10 nM/1 nM in cell-free assays, exhibits >450-fold selectivity over other angiogenic receptors, has antiangiogenic and antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0521184 Supplier Selleck Chemical LLC Supplier No. S15365MG
CP-673451 is a selective inhibitor of PDGFR?/? with IC50 of 10 nM/1 nM in cell-free assays, exhibits >450-fold selectivity over other angiogenic receptors, has antiangiogenic and antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.