Learn More
Apexbio Technology LLC CP-724714, 5mg. Cas: 537705-08-1 MFCD: MFCD15144738. HER2 inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A24125
Encompass_Preferred
CP-724714 is an inhibitor of erbB2 and EGFR kinases with IC50 values of 103 nmol/L and 6,4002,100 nmol/L, respectively [1].In the in vitro cell cycle assay, CP-724714 cause a G1 block of the Her2-amplified BT-474 breast cancer cells due to its inhibition of erbB2. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4742