Learn More
Selleck Chemical LLC CP-724714 5mg 537705-08-1
SDP
Catalog No. 508368
Encompass_Preferred
CP-724714 is a potent, selective inhibitor of HER2/ErbB2 with IC50 of 10 nM, >640-fold selectivity against EGFR, InsR, IRG-1R, PDGFR, VEGFR2, Abl, Src, c-Met etc in cell-free assays. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-836-8 Supplier Selleck Chemical LLC Supplier No. S11675MG
CP-724714 is a potent, selective inhibitor of HER2/ErbB2 with IC50 of 10 nM, >640-fold selectivity against EGFR, InsR, IRG-1R, PDGFR, VEGFR2, Abl, Src, c-Met etc in cell-free assays. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.