Learn More
Cayman Chemical CP 809,101 (hydrochloride), Purity: ≥95%, CAS: 1215721-40-6, 50 mg
SDP
Catalog No. 502949906
Encompass_Preferred
A potent 5-HT2C receptor agonist; selective for human 5-HT2C and rat 5-HT2C over human 5-HT2A, 5-HT2B, and rat 5-HT2A receptors in a FLIPR assay (EC50s = 0.11, 0.06, 153, 65.3, and 119 nM, respectively), as well as human dopamine D2, histamine H1, and α1- and α2-adrenergic receptors (Kis = 872, 828, 217, and 956 nM, respectively); inhibits the conditioned avoidance response, hyperactivity induced by PCP and D-amphetamine, and spontaneous locomotor activity in rats (ED50s = 4.8, 2.4, 2.9, and 2 mg/kg, respectively); reduces reinstatement of drug-seeking behavior in cocaine-primed rats (0.01-1 μg/0.2 μl/side in the central amygdala)
Catalog No. 50-294-9906 Supplier Cayman Chemical Supplier No. 3328350 MG
A potent 5-HT2C receptor agonist; selective for human 5-HT2C and rat 5-HT2C over human 5-HT2A, 5-HT2B, and rat 5-HT2A receptors in a FLIPR assay (EC50s = 0.11, 0.06, 153, 65.3, and 119 nM, respectively), as well as human dopamine D2, histamine H1, and α1- and α2-adrenergic receptors (Kis = 872, 828, 217, and 956 nM, respectively); inhibits the conditioned avoidance response, hyperactivity induced by PCP and D-amphetamine, and spontaneous locomotor activity in rats (ED50s = 4.8, 2.4, 2.9, and 2 mg/kg, respectively); reduces reinstatement of drug-seeking behavior in cocaine-primed rats (0.01-1 μg/0.2 μl/side in the central amygdala)