Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CP 93129 dihydrochloride | 879089-64-2 | 99.0% | 288.17 g·mol⁻1 | C12H15Cl2N3O | 1 MG
SDP

Catalog No. 5000213173
Encompass_Preferred

CP 93129 dihydrochloride is a selective 5-HT1B receptor agonist supplied for research use. It is used in pharmacology and neuroscience studies, including investigations related to Parkinson's disease, and is provided as a high-purity solid suitable for analytical, in vitro, and in vivo applications.

  • Selective 5-HT1B receptor agonist suitable for receptor pharmacology studies.
  • High purity (99.0%) for consistent experimental results.
  • Molecular weight 288.17 g·mol⁻1 and formula C12H15Cl2N3O for precise calculations.
  • Off-white to light yellow solid for convenient handling and storage.
  • Recommended storage at 4°C, sealed and protected from moisture and light.
  • Available in small, accurately weighed milligram quantities for dosing and assays.

Catalog No. 50-002-13173 Supplier Medchemexpress LLC Supplier No. HY101357A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CP 93129 dihydrochloride is a selective 5-HT1B receptor agonist supplied for research use. It is used in pharmacology and neuroscience studies, including investigations related to Parkinson's disease, and is provided as a high-purity solid suitable for analytical, in vitro, and in vivo applications.

  • Selective 5-HT1B receptor agonist suitable for receptor pharmacology studies.
  • High purity (99.0%) for consistent experimental results.
  • Molecular weight 288.17 g·mol⁻1 and formula C12H15Cl2N3O for precise calculations.
  • Off-white to light yellow solid for convenient handling and storage.
  • Recommended storage at 4°C, sealed and protected from moisture and light.
  • Available in small, accurately weighed milligram quantities for dosing and assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.