Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CP 93129 dihydrochloride | 879089-64-2 | MFCD16875420 | 99.0% | 288.17 g/mol | C12H15Cl2N3O | 5 MG
SDP

Catalog No. 5000213151
Encompass_Preferred

CP 93129 dihydrochloride is the dihydrochloride salt of a potent and selective 5-HT1B receptor agonist used as a research reagent in neuroscience, including Parkinson's disease models. Supplied as an off-white to light yellow solid, it is intended for research and analytical applications.

  • Potent, selective 5-HT1B receptor agonist for pharmacological studies.
  • High purity (≈99.0%) suitable for analytical and in vitro work.
  • CAS number 879089-64-2 for unambiguous identification.
  • Molecular formula C12H15Cl2N3O; molecular weight ~288.17 g/mol.
  • Recommended storage: 4°C; in solvent keep at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-13151 Supplier Medchemexpress LLC Supplier No. HY101357A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CP 93129 dihydrochloride is the dihydrochloride salt of a potent and selective 5-HT1B receptor agonist used as a research reagent in neuroscience, including Parkinson's disease models. Supplied as an off-white to light yellow solid, it is intended for research and analytical applications.

  • Potent, selective 5-HT1B receptor agonist for pharmacological studies.
  • High purity (≈99.0%) suitable for analytical and in vitro work.
  • CAS number 879089-64-2 for unambiguous identification.
  • Molecular formula C12H15Cl2N3O; molecular weight ~288.17 g/mol.
  • Recommended storage: 4°C; in solvent keep at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.