Learn More
AVANTOR MASTERFLEX (VWR LLC) Male Luer Adapter, 3/32"", PP, Animal Free, 25/pk
Catalog No. NC0850747
Encompass
Animal Free Male Luer Adapter, 3/32", Polypropylene, 25/pk, Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving).
Catalog No. NC0850747 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3080028
Animal Free Male Luer Adapter, 3/32", Polypropylene, 25/pk, Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving).