Learn More
Cole-Parmer Essentials Sterile Cell Strainers, 40 µm, Blue, Bulk Packed; 50/CS
Catalog No. NC1977153 Shop All Cole-Parmer Products
Encompass_Preferred
Polypropylene housing with nylon mesh filter is a faster, easier alternative to gauze or paper filtration. Elongated lip on housing enable aseptic handling with forceps. Strainers are supplied sterile (gamma irradiated). Choose from 40, 70, or 100 µm mesh pore size strainers. Order convex head pestle with molded textured surface to better manipulate material in the strainer.
Catalog No. NC1977153 Supplier Cole-Parmer Supplier No. 0633665
Polypropylene housing with nylon mesh filter is a faster, easier alternative to gauze or paper filtration. Elongated lip on housing enable aseptic handling with forceps. Strainers are supplied sterile (gamma irradiated). Choose from 40, 70, or 100 µm mesh pore size strainers. Order convex head pestle with molded textured surface to better manipulate material in the strainer.