Learn More
Cole-Parmer Cole-Parmer Compact Digital Mixer System, 50 to 2500 rpm, 115/230 VAC with Plug Adapter Kit
Catalog No. NC1498537 Shop All Cole-Parmer Products
Encompass
This compact, digital mixer is perfect for smaller scale mixing needs throughout your laboratory. The maintenance-free brushless DC motor provides speeds from 50 to 2500 rpm. The LED display lets you make precise speed adjustments and ensures repeatability. The motor housing is lightweight molded nylon, resistant to the occasional splash and spill. Mixer housing includes an integrated stand clamp that allows for quick and secure mounting to the included stand. The stainless steel collet, with set screws, accepts 1/4" (6-mm) shafts.
Catalog No. NC1498537 Supplier Cole-Parmer Supplier No. 5000603
This compact, digital mixer is perfect for smaller scale mixing needs throughout your laboratory. The maintenance-free brushless DC motor provides speeds from 50 to 2500 rpm. The LED display lets you make precise speed adjustments and ensures repeatability. The motor housing is lightweight molded nylon, resistant to the occasional splash and spill. Mixer housing includes an integrated stand clamp that allows for quick and secure mounting to the included stand. The stainless steel collet, with set screws, accepts 1/4" (6-mm) shafts.