Learn More
Lonza Walkersville Human Cryopreserved Hepatocyte Thawing Medium
Catalog No. 501060967
Encompass
Human Cryopreserved Hepatocyte Thawing Medium is supplied ready to use in 50mL conical tubes. Medium has been optimized specifically for single-donor human primary hepatocytes to ensure high viability and yield is maintained during reanimation from cryopreservation.Benefits:Optimized for maintaining hepatocyte cell health during thawingApplications:ADMEToxContent:50mL conical tube
Catalog No. 50-106-0967 Supplier Lonza Walkersville Supplier No. MCHT50
Human Cryopreserved Hepatocyte Thawing Medium is supplied ready to use in 50mL conical tubes. Medium has been optimized specifically for single-donor human primary hepatocytes to ensure high viability and yield is maintained during reanimation from cryopreservation.Benefits:Optimized for maintaining hepatocyte cell health during thawingApplications:ADMEToxContent:50mL conical tube