Learn More
Lonza Walkersville NoSpin HepaRG (8.0M cells/vial)
Catalog No. 501060989
Encompass
HepaRG Cells are fully-functional, adult-phenotype, human hepatic cells. HepaRG Cells exhibit many characteristics similar to primary human hepatocytes and can be used for many of the same applications.The increased metabolic activity of HepaRG Cells compared to HepG2 enables more predictive value of higher throughput liver toxicity assays conducted in early drug discovery.Benefits:Hepatoctye-like morphology and functionWidely accepted as screening tool for liver toxicityNo centrifugation step required for higher throughputGreater metabolic activity than HepG2 cellsApplications:Drug metabolisminfectious diseaseADMEToxContent:1 x Cryopreserved ampule NoSpin HepaRG containing ≥8 million cells
Catalog No. 50-106-0989 Supplier Lonza Walkersville Supplier No. NSHPRG
HepaRG Cells are fully-functional, adult-phenotype, human hepatic cells. HepaRG Cells exhibit many characteristics similar to primary human hepatocytes and can be used for many of the same applications.The increased metabolic activity of HepaRG Cells compared to HepG2 enables more predictive value of higher throughput liver toxicity assays conducted in early drug discovery.Benefits:Hepatoctye-like morphology and functionWidely accepted as screening tool for liver toxicityNo centrifugation step required for higher throughputGreater metabolic activity than HepG2 cellsApplications:Drug metabolisminfectious diseaseADMEToxContent:1 x Cryopreserved ampule NoSpin HepaRG containing ≥8 million cells