Learn More
Cole-Parmer Cole-Parmer Precleaned EPA Sample Jars, 950 mL, 12/cs
Catalog No. NC1397238 Shop All Cole-Parmer Products
Encompass_Preferred
Jars are certified clean for worry-free sampling; comes with a certificate of conformance to EPA Specifications and Guidance for Contaminant-Free Sample Containers. PTFE-lined caps protect contents. Sample analysis labels included (unattached).
Catalog No. NC1397238 Supplier Cole-Parmer Supplier No. 9954044
Jars are certified clean for worry-free sampling; comes with a certificate of conformance to EPA Specifications and Guidance for Contaminant-Free Sample Containers. PTFE-lined caps protect contents. Sample analysis labels included (unattached).