Learn More
Cole-Parmer SH-200D-O-L-120 Large Orbital Shaker; 120 VAC
Supplier:  Cole-Parmer 5190101
Encompass_Preferred
These orbital shakers provide a smooth, uniform circular motion with a 16-mm orbit — ideal for larger samples and multiwell plates. Easily set and view the speed and the timer on the LED display. Control speed from 30 to 300 rpm. The microprocessor maintains speed accuracy even over long runs. Built-in timer can be set from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert. Durable shakers are rated for continuous use. Use in incubators and environmental or cold chambers up to 104°F (40°C).
Catalog No. NC2124226