Learn More
Lonza Walkersville Human Cryopreserved Pooled Donor Hepatocyte Thawing Medium
Catalog No. 501060968
Encompass
Human Cryopreserved Pooled Donor Human Hepatocyte Thawing Medium is supplied ready to use in 50mL conical tubes. Medium has been optimized specifically for DonorPlex pooled donor human primary hepatocytes to ensure high viability and yield is maintained during reanimation from cryopreservation.Benefits:Optimized for thawing DonorPlex HepatocytesReduces dead cells and debris prior to cell useApplications:ADMEToxContent:50mL tubes
Catalog No. 50-106-0968 Supplier Lonza Walkersville Supplier No. MCHT50P
Human Cryopreserved Pooled Donor Human Hepatocyte Thawing Medium is supplied ready to use in 50mL conical tubes. Medium has been optimized specifically for DonorPlex pooled donor human primary hepatocytes to ensure high viability and yield is maintained during reanimation from cryopreservation.Benefits:Optimized for thawing DonorPlex HepatocytesReduces dead cells and debris prior to cell useApplications:ADMEToxContent:50mL tubes