Learn More
Agilent Technologies CP-PORABOND U,
Catalog No. NC2241832
Encompass
CP-PoraBOND U, fused silica, 25 m, 0.32 mm, 7.00 µm
This item is not returnable. View return policy
Catalog No. NC2241832 Supplier Agilent Technologies Supplier No. CP7381
CP-PoraBOND U, fused silica, 25 m, 0.32 mm, 7.00 µm