Learn More
Lonza Walkersville Rodent and Monkey Cryopreserved Hepatocyte Thawing Medium
Catalog No. 501060966
Encompass
Rodent and Monkey Cryopreserved Hepatocyte Thawing Medium is supplied in ready to use in 50mL conical tubes. Medium has been optimized specifically for monkey, mouse, and rat primary hepatocytes to ensure high viability and yield is maintained during reanimation from cryopreservation.Benefits:Optimized for thawing rodent and monkey hepatocytesApplications:ADMEToxContent:50mL conical tube
Catalog No. 50-106-0966 Supplier Lonza Walkersville Supplier No. MCRT50
Rodent and Monkey Cryopreserved Hepatocyte Thawing Medium is supplied in ready to use in 50mL conical tubes. Medium has been optimized specifically for monkey, mouse, and rat primary hepatocytes to ensure high viability and yield is maintained during reanimation from cryopreservation.Benefits:Optimized for thawing rodent and monkey hepatocytesApplications:ADMEToxContent:50mL conical tube