Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cp (Rat) ELISA Kit

Catalog No. 89100692 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-692 Plasma, Serum
89-028-159 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-692 Supplier Abnova™ Supplier No. KA2502
Add to Cart
Add to Cart

Competitive Enzyme Immunoassay for the quantitative measurement of Rat Cp in Plasma and Serum

Specifications

Conjugate HRP
Cross Reactivity Rat
Description Cp (Rat) ELISA Kit is for the quantitative measurement of Cp in plasma and serum.
Detection Limit 4.7 to 300ng/mL
Detection Method Colorimetric
Includes 96 wells Anti mouse ceruloplasmin coated plate, Lyophilized ceruloplasmin stock, 10x Diluent (25mL), Enzyme conjugate reagent (11mL), 20X Wash Solution (50mL),TMB Reagent (11mL), Stop Solution (11mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Cp (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.