Learn More
Cole-Parmer ESS GD0912-7000 Sampling Bags With Combination Valve, 3l
Catalog No. NC0557363 Shop All Cole-Parmer Products
Encompass_Preferred
Gas sampling bags are constructed of 2-mil thick Tedlar with solid seam to keep gases contained. Unique, polypropylene combination valve features 3/16" OD on/off stem suitable for tubing connections and integral PTFE silicone septum for syringe sampling. Low permeability. Select from a variety of sizes to suit your needs. Case of 10.
Catalog No. NC0557363 Supplier Cole-Parmer Supplier No. 8656102
Gas sampling bags are constructed of 2-mil thick Tedlar with solid seam to keep gases contained. Unique, polypropylene combination valve features 3/16" OD on/off stem suitable for tubing connections and integral PTFE silicone septum for syringe sampling. Low permeability. Select from a variety of sizes to suit your needs. Case of 10.