Learn More
ADAPTIVE SURGICAL LLC 493A CP VISORB SUTURE, Size: 5/0. Needle: P-3, 13mm
Catalog No. NC0314307
Encompass
493A CP VISORB SUTURE, Size: 5/0. Needle: P-3, 13mm 3/8C Precision RevCt. Length: 18 in.~45cm. Color: UNDYED, 12/box. (Comparable to Ethicon J493G). Supplier will only ship to the following states: IN, KY, VT, ND, OR, WA, NM, TX, MN, AR, MO, MA, WI, CO, NE, HI, OH.
Catalog No. NC0314307 Supplier ADAPTIVE SURGICAL LLC Supplier No. 493A
493A CP VISORB SUTURE, Size: 5/0. Needle: P-3, 13mm 3/8C Precision RevCt. Length: 18 in.~45cm. Color: UNDYED, 12/box. (Comparable to Ethicon J493G). Supplier will only ship to the following states: IN, KY, VT, ND, OR, WA, NM, TX, MN, AR, MO, MA, WI, CO, NE, HI, OH.