Learn More
Selleck Chemical LLC CP2 5mg
SDP
Supplier:  Selleck Chemical LLC S86015MG
Encompass_Preferred
CP2 is a cyclic peptide that inhibits the JmjC histone demethylases KDM4 with IC50 values of 42 nM and 29 nM for KDM4A and KDM4C, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8164