Learn More
Medchemexpress LLC HY-100207 5mg Medchemexpress, CP21R7 CAS:125314-13-8 Purity:>98%
SDP
Catalog No. 501931750
Encompass_Preferred
Medchemexpress, HY-100207 5mg CP21R7 CAS:125314-13-8 CP21R7 is potent GSK-3β inhibitor, with an IC 50 of 1.8 nM; CP21R7 also shows inhibitory activitiy against PKCα , with an IC 50 of 1900 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1750 Supplier Medchemexpress LLC Supplier No. HY1002075MG
Medchemexpress, HY-100207 5mg CP21R7 CAS:125314-13-8 CP21R7 is potent GSK-3β inhibitor, with an IC 50 of 1.8 nM; CP21R7 also shows inhibitory activitiy against PKCα , with an IC 50 of 1900 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.