Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TechNiGlove CP400 Series Clean Packed Blue Nitrile Gloves
GREENER_CHOICESDP

Catalog No. 19999471 Shop All TechNiGlove International Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-999-471 X-Small
19-999-472 Small
19-999-473 Medium
19-999-474 Large
19-999-475 X-Large
19-999-476 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-999-471 Supplier TechNiGlove Supplier No. CP400
Add to Cart
Add to Cart
This item is not returnable. View return policy

Superior feel and strength versus a typical nitrile exam glove, using new soft modulus technology

  • The new Clean Pack Rival series is the ideal way to clean your lab area, without the expense of a cleanroom glove
  • Superior feel and strength versus a typical nitrile exam glove, using new soft modulus technology
  • The CP400 replaces the cardboard packaging of a boxed exam glove with our double-bagged clean packaging in order to eliminate messy cardboard shedding

Specifications

Chlorinated Yes
Accelerator Free Yes
Applications Industrial, Laboratory
AQL 1.5
Color Purple
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 4 mil
Finger Thickness (Metric) 0.1 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4 mil
Palm Thickness (Metric) 0.1 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Small
Sterility Non-sterile
Thickness (English) 4 mil
Thickness (Metric) 0.1 mm
FDA 510 (k) Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.