Learn More
Medchemexpress LLC HY-130257 5mg Medchemexpress, CP5V CAS:2509359-75-3 Purity:>98%
SDP
Catalog No. 502081233
Encompass_Preferred
Medchemexpress, HY-130257 5mg CP5V CAS:2509359-75-3 CP5V is a PROTAC, which specifically degrades Cdc20 by linking Cdc20 to the VHL/VBC complex for ubiquitination followed by proteasomal degradation. CP5V induces mitotic inhibition and suppresses cancer cell proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1233 Supplier Medchemexpress LLC Supplier No. HY1302575MG
Medchemexpress, HY-130257 5mg CP5V CAS:2509359-75-3 CP5V is a PROTAC, which specifically degrades Cdc20 by linking Cdc20 to the VHL/VBC complex for ubiquitination followed by proteasomal degradation. CP5V induces mitotic inhibition and suppresses cancer cell proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.