Learn More
Medchemexpress LLC CP94253 hydrochloride | 845861-39-4 | MFCD03453293 | 98.8% | 293.79 g·mol⁻¹ | C15H20ClN3O | 1 ML
SDP
Catalog No. 5000246710
Encompass_Preferred
CP94253 hydrochloride is a potent, selective serotonin 5-HT1B receptor agonist supplied for research use and commonly provided as a 10 mM solution in DMSO for in vitro and in vivo studies.
  • Potent 5-HT1B agonist (Ki ≈ 2 nM).
  • High purity (≈98.8%).
  • Supplied as a 10 mM, 1 mL solution in DMSO; also available as solid in multiple milligram quantities.
  • Chemical formula C15H20ClN3O; molecular weight 293.79 g·mol⁻¹.
  • Light yellow solid appearance; store sealed and away from moisture; in solvent: -80°C (up to 6 months) or -20°C (1 month).
Catalog No. 50-002-46710 Supplier Medchemexpress LLC Supplier No. HY10315110MM 1ML
CP94253 hydrochloride is a potent, selective serotonin 5-HT1B receptor agonist supplied for research use and commonly provided as a 10 mM solution in DMSO for in vitro and in vivo studies.
  • Potent 5-HT1B agonist (Ki ≈ 2 nM).
  • High purity (≈98.8%).
  • Supplied as a 10 mM, 1 mL solution in DMSO; also available as solid in multiple milligram quantities.
  • Chemical formula C15H20ClN3O; molecular weight 293.79 g·mol⁻¹.
  • Light yellow solid appearance; store sealed and away from moisture; in solvent: -80°C (up to 6 months) or -20°C (1 month).