Learn More
Medchemexpress LLC CPA inhibitor | 223532-02-3 | 99.78% | C18H19NO4 | 1 ML
SDP
Catalog No. 5000434296
Encompass_Preferred
CPA inhibitor (Compound 5) is a potent carboxypeptidase A (CPA) inhibitor with a Ki of 0.32 μM. It is for research use only and not sold to patients.
  • Potent carboxypeptidase A (CPA) inhibitor
  • Ki of 0.32 μM
  • For research use only
Catalog No. 50-004-34296 Supplier Medchemexpress LLC Supplier No. HY7000510MM/1ML
CPA inhibitor (Compound 5) is a potent carboxypeptidase A (CPA) inhibitor with a Ki of 0.32 μM. It is for research use only and not sold to patients.
  • Potent carboxypeptidase A (CPA) inhibitor
  • Ki of 0.32 μM
  • For research use only