Learn More
Aviva Systems Biology Corp Cpa3 Antibody - N-terminal region (ARP41980_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41980_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cpa3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: MC-CPA, mMC-CPAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Yeast
Catalog No. 50-284-969