Learn More
Aviva Systems Biology Corp CPB2 Antibody - middle region (ARP43570_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43570_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CPB2. Alternate names: CPU, PCPB, TAFIApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58572