Learn More
Grainger Colder Inline Coupler, Chrome Plated
Catalog No. NC2470433
Encompass_Preferred
The Colder Inline Coupler is a chrome-plated brass fitting that connects two pieces of tubing in a straight line. It has barbed ends to grip the tubing and provides a leak-proof seal up to 300 psi. This coupler has a 1/4" NPT female thread on both ends and is suitable for use with air, water, oil, or inert gas. It measures 2-1/2" long x 1" diameter. This durable fitting allows quick connect and disconnect of tubing in industrial, commercial, and residential applications.
Catalog No. NC2470433 Supplier Grainger Supplier No. 2YCU6
The Colder Inline Coupler is a chrome-plated brass fitting that connects two pieces of tubing in a straight line. It has barbed ends to grip the tubing and provides a leak-proof seal up to 300 psi. This coupler has a 1/4" NPT female thread on both ends and is suitable for use with air, water, oil, or inert gas. It measures 2-1/2" long x 1" diameter. This durable fitting allows quick connect and disconnect of tubing in industrial, commercial, and residential applications.