Learn More
Cole-Parmer CPC (Colder) AseptiQuik G Quick-Disconnect Fitting, Genderless Hose Barb, High-Temperature Polycarbonate, 3/4" ID, 1/Ea; Colder AQG17012HT
Catalog No. 5000610264 Shop All Cole-Parmer Products
Encompass_Preferred
Genderless sterilizable connectors simplify process integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. AseptiQuik connectors are made from ADIF materials. High-temperature polycarbonate withstands autoclaving up to 130°C (266°F) for 60 minutes.
    Catalog No. 50-006-10264 Supplier Cole-Parmer Supplier No. 5011161
    Genderless sterilizable connectors simplify process integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. AseptiQuik connectors are made from ADIF materials. High-temperature polycarbonate withstands autoclaving up to 130°C (266°F) for 60 minutes.