Learn More
Cole-Parmer CPC (Colder) AseptiQuik G Quick-Disconnect Fitting, Genderless, Sanitary, Polyphenylsulfone, 3/4" Sanitary, 1/Each; Colder AQG33112
Catalog No. 5000610310 Shop All Cole-Parmer Products
Encompass_Preferred
Genderless sterilizable connectors simplify process integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Polyphenylsulfone (PPSU) offers superior chemical resistance
  • BPA free
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. Connectors are made from ADIF materials.
    Catalog No. 50-006-10310 Supplier Cole-Parmer Supplier No. 5011172
    Genderless sterilizable connectors simplify process integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Polyphenylsulfone (PPSU) offers superior chemical resistance
  • BPA free
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. Connectors are made from ADIF materials.