Learn More
Cole-Parmer CPC (Colder) AseptiQuik L Quick-Disconnect Fitting, High-Flow, Genderless Sanitary Adapter, Polycarbonate, 1-1/2" Sanitary, 1/Each; Colder AQL33024
Catalog No. 5000610258 Shop All Cole-Parmer Products
Encompass_Preferred
High-flow genderless sterilizable connectors simplify process integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Large internal diameter for efficient transfer
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.
    Polycarbonate is resistant to some chemicals, withstands sterilization and is typically transparent.
    Catalog No. 50-006-10258 Supplier Cole-Parmer Supplier No. 5011187
    High-flow genderless sterilizable connectors simplify process integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Large internal diameter for efficient transfer
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.
    Polycarbonate is resistant to some chemicals, withstands sterilization and is typically transparent.