Learn More
Cole-Parmer CPC (Colder) AseptiQuik L Quick-Disconnect Fitting, High-Flow, Genderless Sanitary Adapter, Polyphenylsulfone, 1-1/2" Sanitary, 1/Each; Colder AQL33124
Catalog No. 5000610239 Shop All Cole-Parmer Products
Encompass_Preferred
High-flow genderless connectors simplify process integration
  • Manufactured in an ISO Class 7 certified cleanroom
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Large internal diameter for efficient transfer
  • Polyphenylsulfone (PPSU) offers superior chemical resistance
  • BPA free
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. PPSU is heat and chemical-resistant with high thermal and hydrolytic stability.
    Catalog No. 50-006-10239 Supplier Cole-Parmer Supplier No. 5011189
    High-flow genderless connectors simplify process integration
  • Manufactured in an ISO Class 7 certified cleanroom
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Large internal diameter for efficient transfer
  • Polyphenylsulfone (PPSU) offers superior chemical resistance
  • BPA free
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. PPSU is heat and chemical-resistant with high thermal and hydrolytic stability.