Learn More
Cole-Parmer CPC (Colder) AseptiQuik S Quick-Disconnect Fitting, Low-Flow, Genderless Hose Barb, High-Temperature Polycarbonate, 1/4" ID, 1/Each; Colder AQS17004HT
Catalog No. 5000610257 Shop All Cole-Parmer Products
Encompass_Preferred
Low-flow genderless connectors simplify integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Designed for small-volume fluid transfer
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. Connectors are made from ADIF materials. High-temp PC withstands autoclaving up to 130°C (266°F) for 60 minutes.
    Catalog No. 50-006-10257 Supplier Cole-Parmer Supplier No. 5011193
    Low-flow genderless connectors simplify integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Designed for small-volume fluid transfer
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. Connectors are made from ADIF materials. High-temp PC withstands autoclaving up to 130°C (266°F) for 60 minutes.