Learn More
Cole-Parmer CPC (Colder) AseptiQuik S Quick-Disconnect Fitting, Low-Flow, Genderless Insert, Polycarbonate, MPC insert, 1/Each; Colder AQS17MPC
Catalog No. 5000610333 Shop All Cole-Parmer Products
Encompass_Preferred
Low-flow genderless connectors simplify integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Designed for small-volume fluid transfer
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. Connectors feature an MPC insert that connect to an MPC-series female coupling body.
    Catalog No. 50-006-10333 Supplier Cole-Parmer Supplier No. 5011196
    Low-flow genderless connectors simplify integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, and pull” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Designed for small-volume fluid transfer
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the connectors together and squeeze until you hear an audible “click”, snap the tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves. Connectors feature an MPC insert that connect to an MPC-series female coupling body.