Learn More
Cole-Parmer CPC (Colder) AseptiQuik STC II Quick-Disconnect Fitting, Genderless Sanitary Adapter with AseptiQuik G, High-Temperature Polycarbonate, 3/4" x 1-1/2", 1/Each; Colder AQGSTC2331000HT
Catalog No. 5000610255 Shop All Cole-Parmer Products
Encompass_Preferred
Genderless connectors simplifyintegration while maximizing flexibility
  • Manufactured in an ISO Class 7 cleanroom
  • Sterilize AseptikQuik STC II fitting on stainless steel equipment via SIP
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.
    Unique 3-port design is suitable for a steam-through steam-in-place process, eliminating dead legs and the need for a laminar flow hood. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle.
    Catalog No. 50-006-10255 Supplier Cole-Parmer Supplier No. 5011180
    Genderless connectors simplifyintegration while maximizing flexibility
  • Manufactured in an ISO Class 7 cleanroom
  • Sterilize AseptikQuik STC II fitting on stainless steel equipment via SIP
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.
    Unique 3-port design is suitable for a steam-through steam-in-place process, eliminating dead legs and the need for a laminar flow hood. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle.