Learn More
Cole-Parmer CPC (Colder) AseptiQuik STC II Quick-Disconnect Fitting, Genderless Sanitary Adapter with AseptiQuik G, Polycarbonate, 3/4" x 3/4", 1/Each; Colder AQGSTC2330900
Catalog No. 5000610256 Shop All Cole-Parmer Products
Encompass_Preferred
Genderless connector and Steam-Thru II connector simplify process integration
  • Manufactured in an ISO Class 7 certified cleanroom
  • Sterilize fitting on stainless steel equipment via SIP
  • AseptiQuik connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.
    Unique three-port design is suitable for a true steam-through steam-in-place (SIP) process, eliminating dead legs and the need for a laminar flow hood. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle.
    Catalog No. 50-006-10256 Supplier Cole-Parmer Supplier No. 5011177
    Genderless connector and Steam-Thru II connector simplify process integration
  • Manufactured in an ISO Class 7 certified cleanroom
  • Sterilize fitting on stainless steel equipment via SIP
  • AseptiQuik connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, then pull the tabs to simultaneously remove both membranes from the connector halves.
    Unique three-port design is suitable for a true steam-through steam-in-place (SIP) process, eliminating dead legs and the need for a laminar flow hood. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle.