Learn More
Cole-Parmer CPC (Colder) AseptiQuik STC II Quick-Disconnect Fitting, Genderless Sanitary Adapter with AseptiQuik S, Polycarbonate, 3/4" x 3/4", 1/Each; Colder AQSSTC2330900
Catalog No. 5000610254 Shop All Cole-Parmer Products
Encompass_Preferred
Low-flow connector and Steam-Thru II connector simplify integration
  • Manufactured in an ISO Class 7 certified cleanroom
  • Designed for small-volume fluid transfer
  • Sterilize AseptikQuik STC II fitting on stainless steel equipment via SIP
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap the protective cover, align connectors with the pull tabs, slide the two connectors together and squeeze until you hear an audible “click”, snap the tabs together, and pull the tabs simultaneously to remove membranes.
    Unique 3-port design is suitable for a true steam-through steam-in-place process, eliminating dead legs and the need for a laminar flow hood. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle.
    Catalog No. 50-006-10254 Supplier Cole-Parmer Supplier No. 5011202
    Low-flow connector and Steam-Thru II connector simplify integration
  • Manufactured in an ISO Class 7 certified cleanroom
  • Designed for small-volume fluid transfer
  • Sterilize AseptikQuik STC II fitting on stainless steel equipment via SIP
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap the protective cover, align connectors with the pull tabs, slide the two connectors together and squeeze until you hear an audible “click”, snap the tabs together, and pull the tabs simultaneously to remove membranes.
    Unique 3-port design is suitable for a true steam-through steam-in-place process, eliminating dead legs and the need for a laminar flow hood. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle.