Learn More
Cole-Parmer CPC (Colder) AseptiQuik W Quick-Disconnect Fitting, High-Flow Large-Bore, Genderless Sanitary, High-Temperature Polycarbonate, 1-1/2" Sanitary, 1/Each; Colder AQW33024HT
Catalog No. 5000610253 Shop All Cole-Parmer Products
Encompass_Preferred
High-flow, large-bore genderless sterile connectors simplify process integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom
  • Simple “flip, click, pull, and snap” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, pull the tabs to simultaneously remove both membranes from the connector halves, remove shipping clips, then snap on each side latch. High-temp PC withstands autoclaving up to 130°C (266°F) for 60 minutes.
    Catalog No. 50-006-10253 Supplier Cole-Parmer Supplier No. 5011213
    High-flow, large-bore genderless sterile connectors simplify process integration while maximizing flexibility
  • Manufactured in an ISO Class 7 certified cleanroom
  • Simple “flip, click, pull, and snap” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • AseptiQuik genderless connectors provide secure connections and quick assembly. The interior flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. To use, unsnap and flip down the protective cover, align connectors with the pull tabs, slide the two connectors together and squeeze until you hear an audible “click” from each side, snap the pull tabs together, pull the tabs to simultaneously remove both membranes from the connector halves, remove shipping clips, then snap on each side latch. High-temp PC withstands autoclaving up to 130°C (266°F) for 60 minutes.