Learn More
Cole-Parmer CPC (Colder) AseptiQuik W Quick-Disconnect Fitting, High-Flow Large Bore Genderless Sanitary, Polycarbonate, 1-1/2" Sanitary, 1/Ea; Colder AQW33024
Catalog No. 5000610289 Shop All Cole-Parmer Products
Encompass_Preferred
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, pull, and snap” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Ideal for large-volume, high-flow applications
  • Robust construction provides reliable and repeatable fluid transfer
  • 1.5" nominal flow path size
  • 1-1/2" sanitary connection
  • High-flow, large-bore genderless sterile connectors simplify process integration while maximizing flexibility
    Catalog No. 50-006-10289 Supplier Cole-Parmer Supplier No. 5014366
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Simple “flip, click, pull, and snap” design minimizes operator errors
  • Create quick and easy sterile connections, even in nonsterile environments
  • Ideal for large-volume, high-flow applications
  • Robust construction provides reliable and repeatable fluid transfer
  • 1.5" nominal flow path size
  • 1-1/2" sanitary connection
  • High-flow, large-bore genderless sterile connectors simplify process integration while maximizing flexibility