Learn More
Accuflow Systems CPC HFC Disconnect, Valved Hose Barb, 1/2 in. ID, 10/Pk.
Catalog No. NC4002981
Encompass
This coupling technology is designed for precise fluid management in demanding applications. It is constructed from high-quality polysulfone with an amber tint, meeting USP Class VI and ADCF standards for sensitive applications. The set operates within a temperature range of 34°F to 104°F (1°C to 40°C) and can withstand up to 75 psi (5.17 bar).
  • Intuitive one-step disconnection process
  • Audible 'click' for secure connection
  • Protective thumb latch cover prevents accidental disconnects
  • Laser etched item and lot numbers ensure full traceability
  • Alloy C-276 internal flow path spring
  • Minimizes operational errors and simplifies training
  • No additional equipment required for sterile disconnection
Catalog No. NC4002981 Supplier Accuflow Systems Supplier No. HFCD39SET8HC
This coupling technology is designed for precise fluid management in demanding applications. It is constructed from high-quality polysulfone with an amber tint, meeting USP Class VI and ADCF standards for sensitive applications. The set operates within a temperature range of 34°F to 104°F (1°C to 40°C) and can withstand up to 75 psi (5.17 bar).
  • Intuitive one-step disconnection process
  • Audible 'click' for secure connection
  • Protective thumb latch cover prevents accidental disconnects
  • Laser etched item and lot numbers ensure full traceability
  • Alloy C-276 internal flow path spring
  • Minimizes operational errors and simplifies training
  • No additional equipment required for sterile disconnection