Learn More
CPC™ Polyethylene Drum Inserts
Catalog No. 05113161 Shop All Colder Products Company Products
Change view
Click to view available options
Temp. Range: 29 deg. to 49 deg. C
Catalog No.
05-113-161 Temp. Range: 29 deg. to 49 deg. C
1 options
Catalog No. 05-113-161 Supplier CPC™ Supplier No. DQPRO202BUT
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Specifications
Product Type Drum Insert
Material Polypropylene
Finish Natural
Includes 2 in. American Buttress Thread and EPDM O-Rings
Max. Temperature (Metric) 49°
Pressure Range 0 to 45 psig (0 to 3.1 Bar)