Learn More
TARGETMOL CHEMICALS INC CPCDPK1/TGCDPK1-IN-2 5MG
SDP
Catalog No. 502579242
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CpCDPK1/TgCDPK1-IN-2 is a dual inhibitor of CpCDPK1 and TgCDPK1 with IC50 values of 12 and 5 nM for CpCDPK1 and TgCDPK1 respectively.CpCDPK1/TgCDPK1-IN-2 can be used in the study of diseases associated with infection of toxoplasmas such as Toxoplasma gondii Cryptosporidium parvum and C. hominus. CpCDPK1/TgCDPK1-IN-2 can be used to study diseases associated with infections of toxoplasma gondii (T. gondii) Cryptosporidium parvum (C. parvum) and Cryptosporidium hominus (C. hominus). purity: 99%
Catalog No. 50-257-9242 Supplier TARGETMOL CHEMICALS INC Supplier No. T720815MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CpCDPK1/TgCDPK1-IN-2 is a dual inhibitor of CpCDPK1 and TgCDPK1 with IC50 values of 12 and 5 nM for CpCDPK1 and TgCDPK1 respectively.CpCDPK1/TgCDPK1-IN-2 can be used in the study of diseases associated with infection of toxoplasmas such as Toxoplasma gondii Cryptosporidium parvum and C. hominus. CpCDPK1/TgCDPK1-IN-2 can be used to study diseases associated with infections of toxoplasma gondii (T. gondii) Cryptosporidium parvum (C. parvum) and Cryptosporidium hominus (C. hominus). purity: 99%